Assay Details
| Gene Symbol: MYRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013279.3 | NP_037411.1 | 18 - 19 | 2415 | 60 | |
| XM_005274225.1 | XP_005274282.1 | 18 - 19 | 2503 | 60 | ||
| XM_005274227.1 | XP_005274284.1 | 18 - 19 | 2503 | 60 | ||
| GenBank mRNA | AF086762.1 | - | 18 - 19 | 2415 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

MYRF
Genomic Map