| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145363.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145363.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145363.hg.1 |
TC2000006601.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145363.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145363.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000379070.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000450570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000460006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000467923.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000468817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
ENST00000486875.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006601.hg.1 |
NM_003818.3 |
Refseq |