Assay Details
| Gene Symbol: CAMK2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001222.3 | NP_001213.2 | 14 - 15 | 1167 | 84 | |
| XM_005270199.1 | XP_005270256.1 | 15 - 16 | 1221 | 84 | ||
| XM_005270205.1 | XP_005270262.1 | 15 - 16 | 1221 | 84 | ||
| XM_017016724.1 | XP_016872213.1 | 15 - 16 | 1221 | 84 | ||
| XM_017016727.1 | XP_016872216.1 | 14 - 15 | 1158 | 84 | ||
| GenBank mRNA | L07043.1 | - | 12 - 13 | 842 | 84 | |
| U32509.1 | - | 1 - 2 | 62 | 84 | ||
More Information
Set Membership: |
|

CAMK2G
Genomic Map