| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004717.hg.1 |
ENST00000342756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004717.hg.1 |
ENST00000510663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004717.hg.1 |
ENST00000537129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004717.hg.1 |
NM_006947.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004717.hg.1 |
TC04000332.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
ENST00000342756.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
ENST00000507126.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
ENST00000510663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
NM_001267722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
NM_006947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
ENST00000342756.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
ENST00000507126.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
ENST00000510663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
NM_001267722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
NM_006947.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151965.hg.1 |
TC0400007593.hg.1 |
Clariom D TC |