Assay Details
| Gene Symbol: TGM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254734.1 | NP_001241663.1 | 8 - 9 | 1155 | 83 | |
| NM_198994.2 | NP_945345.2 | 8 - 9 | 1155 | 83 | ||
| GenBank mRNA | AF540969.1 | - | 8 - 9 | 1155 | 83 | |
| AF540970.1 | - | 8 - 9 | 1155 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000024.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20000298.hg.1 | ENST00000202625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000298.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000298.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000298.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000298.hg.1 | TC20000024.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000477505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000477505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000477505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | ENST00000477505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000297.hg.1 | ENST00000202625.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000297.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000297.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000297.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000297.hg.1 | TC20000024.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000477505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000477505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000477505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | ENST00000477505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006510.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | TC2000006510.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144620.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | NM_198994.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | TC2000006510.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | ENST00000202625.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | ENST00000381423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | NM_001254734.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144621.hg.1 | NM_198994.2 | Refseq |

TGM6
Genomic Map