Assay Details
| Gene Symbol: AKR1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190906.1 | NP_001177835.1 | 6 - 7 | 816 | 137 | |
| NM_005989.3 | NP_005980.1 | 7 - 8 | 939 | 137 | ||
| GenBank mRNA | AK058142.1 | - | 8 - 9 | 875 | 137 | |
| AK289425.1 | - | 7 - 8 | 879 | 137 | ||
| AK298428.1 | - | 6 - 7 | 782 | 137 | ||
| BC130625.1 | - | 7 - 8 | 894 | 40146656 | 137 | |
| BC130627.1 | - | 7 - 8 | 894 | 40146658 | 137 | |
| Z28339.1 | - | 7 - 8 | 921 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000858.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
bile acid biosynthetic process
cholesterol catabolic process
digestion
C21-steroid hormone metabolic process
androgen metabolic process
bile acid catabolic process
oxidation-reduction process
aldo-keto reductase (NADP) activity
steroid binding
delta4-3-oxosteroid 5beta-reductase activity
bile acid biosynthetic process
cholesterol catabolic process
digestion
C21-steroid hormone metabolic process
androgen metabolic process
bile acid catabolic process
oxidation-reduction process
aldo-keto reductase (NADP) activity
steroid binding
delta4-3-oxosteroid 5beta-reductase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000438242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000438242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000438242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000438242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000470851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000470851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | ENST00000470851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013455.hg.1 | ENST00000242375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013455.hg.1 | ENST00000297463.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013455.hg.1 | ENST00000411726.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013455.hg.1 | ENST00000468877.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013455.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013455.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013455.hg.1 | TC07000858.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198966.hg.1 | TC0700013466.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000242375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000411726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000432161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000438242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000438242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000438242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000438242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000470851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000470851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | ENST00000470851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_001190907.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013466.hg.1 | NM_005989.3 | Refseq |

AKR1D1
Genomic Map