Assay Details
| Gene Symbol: APOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001647.3 | NP_001638.1 | 2 - 3 | 492 | 78 | |
| GenBank mRNA | AK294523.1 | - | 2 - 3 | 308 | 78 | |
| AK312090.1 | - | 2 - 3 | 227 | 78 | ||
| BC007402.2 | - | 2 - 3 | 207 | 3050012 | 78 | |
| BT019860.1 | - | 1 - 2 | 130 | 78 | ||
| BT019861.1 | - | 1 - 2 | 130 | 78 | ||
| BU195270.1 | - | 2 - 3 | 405 | 78 | ||
| CR456838.1 | - | 1 - 2 | 130 | 78 | ||
| CR541773.1 | - | 1 - 2 | 130 | 78 | ||
| DA892855.1 | - | 2 - 3 | 78 | |||
| J02611.1 | - | 2 - 3 | 191 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002146.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
angiogenesis
glucose metabolic process
lipid metabolic process
lipid transport
brain development
aging
negative regulation of platelet-derived growth factor receptor signaling pathway
peripheral nervous system axon regeneration
tissue regeneration
negative regulation of protein import into nucleus
response to drug
negative regulation of smooth muscle cell proliferation
response to axon injury
negative regulation of focal adhesion assembly
negative regulation of lipoprotein lipid oxidation
negative regulation of monocyte chemotactic protein-1 production
negative regulation of cytokine production involved in inflammatory response
negative regulation of smooth muscle cell-matrix adhesion
negative regulation of T cell migration
lipid transporter activity
protein binding
cholesterol binding
response to reactive oxygen species
angiogenesis
glucose metabolic process
lipid metabolic process
lipid transport
brain development
aging
negative regulation of platelet-derived growth factor receptor signaling pathway
peripheral nervous system axon regeneration
tissue regeneration
negative regulation of protein import into nucleus
response to drug
negative regulation of smooth muscle cell proliferation
response to axon injury
negative regulation of focal adhesion assembly
negative regulation of lipoprotein lipid oxidation
negative regulation of monocyte chemotactic protein-1 production
negative regulation of cytokine production involved in inflammatory response
negative regulation of smooth muscle cell-matrix adhesion
negative regulation of T cell migration
lipid transporter activity
protein binding
cholesterol binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037894.hg.1 | ENST00000343267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037894.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037894.hg.1 | TC03002146.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037885.hg.1 | ENST00000343267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037885.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037885.hg.1 | TC03002146.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037887.hg.1 | ENST00000343267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037887.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037887.hg.1 | TC03002146.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037890.hg.1 | ENST00000343267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037890.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037890.hg.1 | TC03002146.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037893.hg.1 | ENST00000343267.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037893.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037893.hg.1 | TC03002146.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013645.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201839.hg.1 | TC0300013645.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201837.hg.1 | TC0300013645.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201843.hg.1 | TC0300013645.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000421243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000453131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000458447.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201840.hg.1 | TC0300013645.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300201847.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201847.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201847.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201847.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201847.hg.1 | TC0300013645.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300201846.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201846.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201846.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201846.hg.1 | ENST00000343267.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201846.hg.1 | ENST00000463719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201846.hg.1 | NM_001647.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201846.hg.1 | TC0300013645.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 127530 | - | - |

APOD
Genomic Map