Assay Details
| Gene Symbol: RPAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024813.2 | NP_079089.2 | 11 - 12 | 1803 | 94 | |
| GenBank mRNA | AK023212.1 | - | 11 - 12 | 1720 | 94 | |
| AK225054.1 | - | 11 - 12 | 1715 | 94 | ||
| BC039014.1 | - | 11 - 12 | 1803 | 5744281 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000853.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01013563.hg.1 | ENST00000370343.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013563.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013563.hg.1 | TC01000853.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164467.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164467.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164467.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164467.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164467.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164467.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164467.hg.1 | TC0100009040.hg.1 | Clariom D TC |

RPAP2
Genomic Map