Assay Details
| Gene Symbol: RPAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024813.2 | NP_079089.2 | 12 - 13 | 1938 | 148 | |
| GenBank mRNA | AK023212.1 | - | 12 - 13 | 1855 | 148 | |
| BC031070.1 | - | 13 - 14 | 1906 | 5313284 | 148 | |
| BC039014.1 | - | 12 - 13 | 1938 | 5744281 | 148 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000853.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01013565.hg.1 | ENST00000370343.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013565.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013565.hg.1 | TC01000853.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013566.hg.1 | ENST00000370343.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013566.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013566.hg.1 | TC01000853.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013564.hg.1 | ENST00000370343.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013564.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013564.hg.1 | TC01000853.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100164470.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164470.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164470.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164470.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164470.hg.1 | TC0100009040.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100164471.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164471.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164471.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164471.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164471.hg.1 | TC0100009040.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000484158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009040.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164469.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164469.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164469.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164469.hg.1 | ENST00000477322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164469.hg.1 | ENST00000610020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164469.hg.1 | NM_024813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164469.hg.1 | TC0100009040.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 176204 | - | - |

RPAP2
Genomic Map