| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011315.hg.1 |
ENST00000603626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011315.hg.1 |
ENST00000603626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011315.hg.1 |
ENST00000603626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013555.hg.1 |
ENST00000370343.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013555.hg.1 |
ENST00000394482.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013555.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013555.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01013555.hg.1 |
TC01000853.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100164457.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164457.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164457.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100164457.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164457.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100164457.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100164457.hg.1 |
TC0100009040.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000477322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000484158.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
ENST00000610020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009040.hg.1 |
NM_024813.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011315.hg.1 |
ENST00000603626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011315.hg.1 |
ENST00000603626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011315.hg.1 |
ENST00000603626.1 |
Ensembl |