Assay Details
| Gene Symbol: ARL13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162491.1 | NP_001155963.1 | 1 - 2 | 104 | 109 | |
| NR_103708.1 | 1 - 2 | 104 | 109 | |||
| NR_103709.1 | 1 - 2 | 104 | 109 | |||
| XM_011530950.2 | XP_011529252.1 | 1 - 2 | 801 | 109 | ||
| XM_011530951.2 | XP_011529253.1 | 1 - 2 | 801 | 109 | ||
| GenBank mRNA | AK301813.1 | - | 1 - 2 | 104 | 109 | |
| BC140808.1 | - | 1 - 2 | 178 | 9021676 | 109 | |
| BC144603.1 | - | 1 - 2 | 178 | 9053134 | 109 | |
| DB727708.1 | - | 1 - 2 | 101 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000471.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK301813.1,BC140808.1,BC144603.1,NM_001162491.1,XM_011530950.2,XM_011530951.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006081.hg.1 | ENST00000450049.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006081.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006081.hg.1 | TC0X000471.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006080.hg.1 | ENST00000450049.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006080.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006080.hg.1 | TC0X000471.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153819.hg.1 | TC0X00007930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153821.hg.1 | TC0X00007930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153823.hg.1 | TC0X00007930.hg.1 | Clariom D TC |

ARL13A
Genomic Map