Assay Details
| Gene Symbol: ARL13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162491.1 | NP_001155963.1 | 5 - 6 | 597 | 110 | |
| NR_103708.1 | 5 - 6 | 597 | 110 | |||
| NR_103709.1 | 5 - 6 | 597 | 110 | |||
| XM_011530950.2 | XP_011529252.1 | 5 - 6 | 1294 | 110 | ||
| GenBank mRNA | AK301813.1 | - | 5 - 6 | 597 | 110 | |
| BC140808.1 | - | 5 - 6 | 671 | 9021676 | 110 | |
| BC144603.1 | - | 5 - 6 | 671 | 9053134 | 110 | |
| CD300373.1 | - | 2 - 3 | 321 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000471.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006085.hg.1 | ENST00000450049.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006085.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006085.hg.1 | TC0X000471.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153828.hg.1 | TC0X00007930.hg.1 | Clariom D TC |

ARL13A
Genomic Map