Assay Details
| Gene Symbol: ARL13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103708.1 | 7 - 8 | 857 | 119 | ||
| NR_103709.1 | 7 - 8 | 857 | 119 | |||
| GenBank mRNA | AK301813.1 | - | 7 - 8 | 857 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000372953.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450049.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NM_001162491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007930.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | ENST00000494863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | NR_103708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153831.hg.1 | TC0X00007930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00153832.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153832.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153832.hg.1 | ENST00000450457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153832.hg.1 | NR_103709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153832.hg.1 | TC0X00007930.hg.1 | Clariom D TC |

ARL13A
Genomic Map