| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004256.hg.1 |
ENST00000358077.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004256.hg.1 |
ENST00000408954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004256.hg.1 |
ENST00000469640.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004256.hg.1 |
ENST00000472284.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004256.hg.1 |
ENST00000491893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004256.hg.1 |
NM_004938.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004256.hg.1 |
TC09000398.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000358077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000408954.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000463069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000469067.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000469640.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000472284.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000489291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000491893.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000622514.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
NM_001288729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
NM_001288730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
NM_001288731.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
NM_004938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000358077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000408954.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000463069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000469067.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000469640.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000472284.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000489291.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000491893.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
ENST00000622514.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
NM_001288729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
NM_001288730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
NM_001288731.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
NM_004938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900152700.hg.1 |
TC0900007807.hg.1 |
Clariom D TC |