Assay Details
| Gene Symbol: STKLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153710.4 | NP_714921.4 | 6 - 7 | 529 | 68 | |
| GenBank mRNA | BC036504.2 | - | 6 - 7 | 529 | 5265506 | 68 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000774.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09009962.hg.1 | ENST00000371955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09009962.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09009962.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09009962.hg.1 | TC09000774.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000462310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000462310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000462310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000462310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000468046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000468046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000468046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000475232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000475232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000475232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000477284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000477284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000477284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | ENST00000477284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900161779.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900161779.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900161779.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900161779.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900161779.hg.1 | TC0900009076.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000371957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000462310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000462310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000462310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000462310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000468046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000468046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000468046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000475232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000475232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000475232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000477284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000477284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000477284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | ENST00000477284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NM_153710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009076.hg.1 | NR_103997.1 | Refseq |

STKLD1
Genomic Map