Assay Details
| Gene Symbol: LINC01210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109987.1 | 3 - 4 | 385 | 100 | ||
| GenBank mRNA | BC038725.2 | - | 2 - 3 | 165 | 100 | |
| BI460052.1 | - | 3 - 4 | 200 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008929.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000465283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000469216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000471091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | ENST00000478772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | NR_109987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163888.hg.1 | TC0300008929.hg.1 | Clariom D TC |

LINC01210
Genomic Map