| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000713.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000713.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000713.hg.1 |
TC06000073.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000712.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000712.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000712.hg.1 |
TC06000073.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600147339.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147339.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147339.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147339.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147339.hg.1 |
TC0600006926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600147341.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147341.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147341.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147341.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147341.hg.1 |
TC0600006926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
ENST00000379568.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006926.hg.1 |
NM_017906.2 |
Refseq |