Assay Details
| Gene Symbol: PPP1R1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261424.1 | NP_001248353.1 | 1 - 2 | 368 | 125 | |
| GenBank mRNA | AF494535.1 | - | 1 - 2 | 315 | 125 | |
More Information
Set Membership: |
|

PPP1R1C
Genomic Map