| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17021203.hg.1 |
ENST00000262418.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17021203.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17021203.hg.1 |
TC17001577.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000471005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000471005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000471005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000471005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000631130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
ENST00000631130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700186543.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700186543.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700186543.hg.1 |
TC1700010820.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700186543.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700186543.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000262418.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000399246.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000471005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000471005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000471005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000471005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000497360.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000498270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000631130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
ENST00000631130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010820.hg.1 |
NM_000342.3 |
Refseq |