Assay Details
| Gene Symbol: CDH15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004933.2 | NP_004924.1 | 6 - 7 | 868 | 92 | |
| GenBank mRNA | BC008951.2 | - | 6 - 7 | 843 | 3027886 | 92 |
| D83542.1 | - | 6 - 7 | 868 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000692.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009764.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009764.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009764.hg.1 | TC16000692.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009765.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009765.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009765.hg.1 | TC16000692.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000521087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | ENST00000524089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008868.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600166161.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600166161.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600166161.hg.1 | TC1600008868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600166161.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600166161.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600166160.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600166160.hg.1 | NM_004933.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600166160.hg.1 | TC1600008868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600166160.hg.1 | ENST00000289746.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600166160.hg.1 | NM_004933.2 | Refseq |

CDH15
Genomic Map