| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013139.hg.1 |
ENST00000389467.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013139.hg.1 |
ENST00000446231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013139.hg.1 |
NM_015092.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013139.hg.1 |
TC16000916.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600173594.hg.1 |
ENST00000446231.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600173594.hg.1 |
ENST00000565324.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600173594.hg.1 |
NM_015092.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600173594.hg.1 |
TC1600009620.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600173594.hg.1 |
ENST00000446231.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600173594.hg.1 |
ENST00000565324.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600173594.hg.1 |
NM_015092.4 |
Refseq |