Assay Details
| Gene Symbol: OXNAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005265559.3 | XP_005265616.1 | 9 - 10 | 1452 | 64 | |
| XM_017007486.1 | XP_016862975.1 | 9 - 10 | 1467 | 64 | ||
More Information
Set Membership: |
|

OXNAD1
Genomic Map