| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030701.hg.1 |
ENST00000340645.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030701.hg.1 |
ENST00000393667.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030701.hg.1 |
NM_004487.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030701.hg.1 |
TC03001698.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
ENST00000340645.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
ENST00000393667.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
ENST00000482512.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
ENST00000614479.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
NM_001256486.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
NM_001256487.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
NM_001256488.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
NM_004487.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
ENST00000340645.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
ENST00000393667.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
ENST00000482512.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
ENST00000614479.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
NM_001256486.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
NM_001256487.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
NM_001256488.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
NM_004487.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190632.hg.1 |
TC0300012170.hg.1 |
Clariom D TC |