Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PR domain 12 | PRDM12 |
HSAN8, PFM9 |
-14.8kb | Anti-sense | Chr. 9 - 130664594 - 130682997 on NCBI Build GRCh38 |
| Downstream |
ABL proto-oncogene 1, non-receptor tyrosine kinase |
ABL1 |
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl |
14.0kb | Anti-sense | Chr. 9 - 130713881 - 130887675 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map