Assay Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297761.1 | NP_001284690.1 | 5 - 6 | 1083 | 59 | |
| NM_001297762.1 | NP_001284691.1 | 5 - 6 | 1220 | 59 | ||
| NM_018208.3 | NP_060678.2 | 6 - 7 | 1343 | 59 | ||
| GenBank mRNA | AB073608.1 | - | 6 - 7 | 943 | 59 | |
| AK293426.1 | - | 5 - 6 | 791 | 59 | ||
| AK302145.1 | - | 5 - 6 | 820 | 59 | ||
| AK316493.1 | - | 5 - 6 | 579 | 59 | ||
| BC010082.2 | - | 6 - 7 | 1164 | 4329734 | 59 | |
More Information
Set Membership: |
|

ETNK2
Genomic Map