| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003583.hg.1 |
ENST00000308059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003583.hg.1 |
ENST00000346219.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003583.hg.1 |
ENST00000452631.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003583.hg.1 |
NM_007335.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003583.hg.1 |
NM_007337.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003583.hg.1 |
TC03000186.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
ENST00000308059.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
ENST00000346219.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
ENST00000478428.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
NM_007335.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
NM_007337.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
ENST00000308059.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
ENST00000346219.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
ENST00000478428.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
NM_007335.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
NM_007337.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149381.hg.1 |
TC0300007065.hg.1 |
Clariom D TC |