| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025275.hg.1 |
ENST00000349062.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025275.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025275.hg.1 |
ENST00000355947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025275.hg.1 |
ENST00000542799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025275.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025275.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025275.hg.1 |
TC07003367.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025274.hg.1 |
ENST00000349062.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025274.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025274.hg.1 |
ENST00000355947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025274.hg.1 |
ENST00000542799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025274.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025274.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07025274.hg.1 |
TC07003367.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013594.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201316.hg.1 |
TC0700013594.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
ENST00000349062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
ENST00000354241.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
ENST00000355947.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
ENST00000496896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
NM_033017.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
NM_033091.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700201317.hg.1 |
TC0700013594.hg.1 |
Clariom D TC |