Assay Details
| Gene Symbol: RBMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016024.2 | NP_057108.2 | 1 - 2 | 64 | 75 | |
| GenBank mRNA | AK292855.1 | - | 1 - 2 | 46 | 75 | |
| AL050405.1 | - | 1 - 2 | 39 | 75 | ||
| BC003676.1 | - | 1 - 2 | 64 | 75 | ||
| BC105582.1 | - | 1 - 2 | 51 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000623.hg
Additional Information:
This assay is in the 5'UTR region of the AK292855.1,AL050405.1,BC003676.1,BC105582.1,NM_016024.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011269.hg.1 | ENST00000424312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015450.hg.1 | ENST00000450122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008251.hg.1 | ENST00000305536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008251.hg.1 | ENST00000538614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008251.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008251.hg.1 | TC0X000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010992.hg.1 | ENST00000257020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010992.hg.1 | ENST00000257020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008255.hg.1 | ENST00000305536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008255.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008255.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008255.hg.1 | ENST00000538614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008255.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008255.hg.1 | TC0X000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000487274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000487274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008253.hg.1 | ENST00000305536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008253.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008253.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008253.hg.1 | ENST00000538614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008253.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008253.hg.1 | TC0X000623.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500011269.hg.1 | ENST00000424312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000487274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000487274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008405.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015450.hg.1 | ENST00000450122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010992.hg.1 | ENST00000257020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010992.hg.1 | ENST00000257020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157263.hg.1 | TC0X00008405.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | ENST00000370947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | ENST00000469953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157266.hg.1 | TC0X00008405.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00157261.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157261.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157261.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157261.hg.1 | ENST00000305536.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157261.hg.1 | ENST00000619267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157261.hg.1 | NM_016024.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157261.hg.1 | TC0X00008405.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 139091 | - | - |

RBMX2
Genomic Map