| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000370947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000370947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000370947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000370947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000487274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000487274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008258.hg.1 |
ENST00000305536.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008258.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008258.hg.1 |
ENST00000538614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008258.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008258.hg.1 |
TC0X000623.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000370947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000370947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000370947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000370947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000487274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000487274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008405.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
ENST00000305536.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
ENST00000469953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
ENST00000619267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
NM_016024.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00157269.hg.1 |
TC0X00008405.hg.1 |
Clariom D TC |