| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000430742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000430742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000430742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000430742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000488728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000488728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000488728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
ENST00000488728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005921.hg.1 |
ENST00000263657.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005921.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005921.hg.1 |
TC02000395.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200154428.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154428.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154428.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154428.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154428.hg.1 |
TC0200007886.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000263657.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000430742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000430742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000430742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000430742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000488728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000488728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000488728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
ENST00000488728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007886.hg.1 |
NM_020143.3 |
Refseq |