Assay Details
| Gene Symbol: LCORL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166139.2 | NP_001159611.1 | 6 - 7 | 901 | 83 | |
| XM_017007964.1 | XP_016863453.1 | 7 - 8 | 777 | 83 | ||
| GenBank mRNA | AL700364.1 | - | 1 - 2 | 89 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001070.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014738.hg.1 | ENST00000382224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014738.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014738.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014738.hg.1 | TC04001070.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400169714.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169714.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169714.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169714.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169714.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169714.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169714.hg.1 | TC0400010168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000326877.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000382226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000510451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | ENST00000512376.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_001166139.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010168.hg.1 | NM_153686.8 | Refseq |

LCORL
Genomic Map