Assay Details
| Gene Symbol: RAB41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530948.2 | XP_011529250.1 | 1 - 2 | 1360 | 79 | |
| XM_017029488.1 | XP_016884977.1 | 1 - 2 | 139 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000276066.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000374473.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | ENST00000509895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007536.hg.1 | NM_001032726.2 | Refseq |

RAB41
Genomic Map