Assay Details
| Gene Symbol: CATSPERB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK024360.1 | - | 1 - 2 | 119 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK024360.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |

CATSPERB
Genomic Map