| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
ENST00000283977.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
ENST00000506587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
ENST00000512866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
ENST00000513973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
NM_001199917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
NM_001199918.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
NM_001199919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
NM_015599.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023232.hg.1 |
TC06001911.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
ENST00000283977.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
ENST00000506587.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
ENST00000512866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
ENST00000513973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
NM_001199917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
NM_001199918.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
NM_001199919.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
NM_015599.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023231.hg.1 |
TC06001911.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000283977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000506587.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000509219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000512866.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000513973.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000616566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
NM_001199917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
NM_001199918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
NM_001199919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
NM_015599.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000283977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000506587.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000509219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000512866.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000513973.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
ENST00000616566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
NM_001199917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
NM_001199918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
NM_001199919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
NM_015599.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185587.hg.1 |
TC0600012432.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000283977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000506587.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000507404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000509219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000512866.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000513973.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000616566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
NM_001199917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
NM_001199918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
NM_001199919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
NM_015599.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000283977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000506587.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000507404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000509219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000512866.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000513973.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
ENST00000616566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
NM_001199917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
NM_001199918.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
NM_001199919.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
NM_015599.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185588.hg.1 |
TC0600012432.hg.1 |
Clariom D TC |