| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060655.hg.1 |
ENST00000366581.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060655.hg.1 |
ENST00000366582.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060655.hg.1 |
NM_018072.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060655.hg.1 |
TC01004007.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100234789.hg.1 |
ENST00000366581.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234789.hg.1 |
ENST00000366582.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234789.hg.1 |
NM_018072.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234789.hg.1 |
ENST00000366581.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234789.hg.1 |
ENST00000366582.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234789.hg.1 |
NM_018072.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234789.hg.1 |
TC0100017860.hg.1 |
Clariom D TC |