Assay Details
| Gene Symbol: CHPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284295.1 | NP_001271224.1 | 1 - 2 | 330 | 81 | |
| GenBank mRNA | AK056331.1 | - | 1 - 2 | 330 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000390183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000465601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000465601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000465601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000465601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009696.hg.1 | NR_030407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169203.hg.1 | TC0700009696.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169204.hg.1 | TC0700009696.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000390183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000465601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000465601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000465601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000465601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000482173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009696.hg.1 | NR_030407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | ENST00000035307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | ENST00000495645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | NM_001284295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | NM_019015.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700169208.hg.1 | TC0700009696.hg.1 | Clariom D TC |

CHPF2
Genomic Map