| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300150983.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300150983.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300150983.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300150983.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300150983.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300150983.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300150983.hg.1 |
TC1300007546.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007546.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300150982.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300150982.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300150982.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300150982.hg.1 |
ENST00000377462.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300150982.hg.1 |
ENST00000449753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300150982.hg.1 |
NM_001258406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300150982.hg.1 |
TC1300007546.hg.1 |
Clariom D TC |