Assay Details
| Gene Symbol: WDR86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006715966.2 | XP_006716029.1 | 5 | 1990 | 65 | |
| GenBank mRNA | AK096225.1 | - | 1 | 953 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | negative regulator of ubiquitin like proteins 1 | NUB1 |
BS4, NUB1L, NYREN18 |
-5.5kb | Sense | Chr. 7 - 151341699 - 151378461 on NCBI Build GRCh38 |
| Downstream | WDR86 antisense RNA 1 | WDR86-AS1 | 24.8kb | Sense | Chr. 7 - 151409161 - 151413354 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

WDR86
Genomic Map