Assay Details
| Gene Symbol: DEDD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039712.1 | NP_001034801.1 | 2 - 3 | 112 | 90 | |
| XM_005245599.2 | XP_005245656.1 | 2 - 3 | 128 | 90 | ||
| GenBank mRNA | BG565440.1 | - | 2 - 3 | 113 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003417.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001039712.1,XM_005245599.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
spermatogenesis
extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of protein catabolic process
regulation of apoptotic process
decidualization
negative regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
DNA binding
protein binding
transcription, DNA-templated
spermatogenesis
extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of protein catabolic process
regulation of apoptotic process
decidualization
negative regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | ENST00000368005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | ENST00000368006.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | ENST00000392188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | ENST00000458050.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | ENST00000535389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | ENST00000541906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | ENST00000545495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053523.hg.1 | TC01003417.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | ENST00000368005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | ENST00000368006.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | ENST00000392188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | ENST00000458050.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | ENST00000535389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | ENST00000541906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | ENST00000545495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053524.hg.1 | TC01003417.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | ENST00000368005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | ENST00000368006.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | ENST00000392188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | ENST00000458050.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | ENST00000541906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | ENST00000545495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053522.hg.1 | TC01003417.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000368005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000368006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000458050.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000463227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000464113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000490843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000545495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000368005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000368006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000458050.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000463227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000464113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000490843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | ENST00000545495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223010.hg.1 | TC0100016159.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000368005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000368006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000458050.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000463227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000464113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000473679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000490843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000545495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000368005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000368006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000458050.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000463227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000464113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000473679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000490843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | ENST00000545495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223011.hg.1 | TC0100016159.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000368005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000368006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000458050.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000463227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000464113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000473679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000486041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000490843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000496632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000545495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000368005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000368006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000458050.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000463227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000464113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000473679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000486041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000490843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000496632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | ENST00000545495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | NM_001039711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | NM_001039712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | NM_032998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223013.hg.1 | TC0100016159.hg.1 | Clariom D TC |

DEDD
Genomic Map