Assay Details
| Gene Symbol: ATP8B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020452.3 | NP_065185.1 | 14 - 15 | 1390 | 65 | |
| XM_005245355.2 | XP_005245412.1 | 13 - 14 | 1537 | 65 | ||
| XM_005245356.3 | XP_005245413.1 | 14 - 15 | 1490 | 65 | ||
| XM_017001871.1 | XP_016857360.1 | 13 - 14 | 1537 | 65 | ||
| XM_017001872.1 | XP_016857361.1 | 14 - 15 | 1486 | 65 | ||
| GenBank mRNA | AB032963.1 | - | 4 - 5 | 522 | 65 | |
| AY302537.1 | - | 14 - 15 | 1390 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001274.hg.1
Set Membership: |
|

ATP8B2
Genomic Map