Assay Details
| Gene Symbol: HSD3B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142778.1 | NP_001136250.1 | 1 - 2 | 99 | 55 | |
| NM_025193.3 | NP_079469.2 | 1 - 2 | 99 | 55 | ||
| GenBank mRNA | AK290950.1 | - | 1 - 2 | 53 | 55 | |
| BC004929.1 | - | 1 - 2 | 86 | 3626629 | 55 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000365.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290950.1,BC004929.1,NM_001142778.1,NM_025193.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16005318.hg.1 | ENST00000262520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005318.hg.1 | ENST00000297679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005318.hg.1 | ENST00000353250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005318.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005318.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005318.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005318.hg.1 | TC16000365.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005320.hg.1 | ENST00000262520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005320.hg.1 | ENST00000297679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005320.hg.1 | ENST00000353250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005320.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005320.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005320.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005320.hg.1 | TC16000365.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007505.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | TC1600007505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154889.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | NM_025193.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | TC1600007505.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | ENST00000262520.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | ENST00000297679.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | ENST00000562932.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | ENST00000574447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | NM_001142777.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | NM_001142778.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154885.hg.1 | NM_025193.3 | Refseq |

HSD3B7
Genomic Map