Assay Details
| Gene Symbol: RPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278283.1 | NP_001265212.1 | 3 - 4 | 333 | 75 | |
| NM_001278285.1 | NP_001265214.1 | 2 - 3 | 302 | 75 | ||
| NM_001278289.1 | NP_001265218.1 | 2 - 3 | 302 | 75 | ||
| NM_001318926.1 | NP_001305855.1 | 2 - 3 | 302 | 75 | ||
| NM_001318927.1 | NP_001305856.1 | 2 - 3 | 302 | 75 | ||
| NM_001318928.1 | NP_001305857.1 | 2 - 3 | 302 | 75 | ||
| NM_001318929.1 | NP_001305858.1 | 2 - 3 | 302 | 75 | ||
| NM_001318930.1 | NP_001305859.1 | 2 - 3 | 291 | 75 | ||
| NM_199229.2 | NP_954699.1 | 2 - 3 | 302 | 75 | ||
| XM_006712677.3 | XP_006712740.1 | 3 - 4 | 249 | 75 | ||
| GenBank mRNA | AK093658.1 | - | 3 - 4 | 264 | 75 | |
| AK291035.1 | - | 2 - 3 | 229 | 75 | ||
| AK303184.1 | - | 2 - 3 | 229 | 75 | ||
| AL833120.1 | - | 2 - 3 | 225 | 75 | ||
| BC005148.2 | - | 2 - 3 | 210 | 3505010 | 75 | |
| BC016764.1 | - | 2 - 3 | 239 | 4107315 | 75 | |
| BC072401.1 | - | 2 - 3 | 211 | 6047438 | 75 | |
| BG546879.1 | - | 2 - 3 | 240 | 75 | ||
| BX403091.2 | - | 2 - 3 | 208 | 75 | ||
| BX643719.1 | - | 2 - 3 | 228 | 75 | ||
| CN372165.1 | - | 2 - 3 | 225 | 75 | ||
| CR992340.1 | - | 2 - 3 | 202 | 75 | ||
| DA251418.1 | - | 1 | 213 | 75 | ||
| DA592494.1 | - | 2 - 3 | 230 | 75 | ||
| DC403549.1 | - | 2 - 3 | 302 | 75 | ||
| Gene Symbol: RPEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143909.1 | NP_001137381.1 | 1 | 315 | 75 | |
| GenBank mRNA | BC137242.1 | - | 1 | 254 | 9021239 | 75 |
| BC137243.1 | - | 1 | 254 | 9021240 | 75 | |
| BC157829.1 | - | 1 | 254 | 9057048 | 75 | |
| BC157875.1 | - | 1 | 316 | 9057093 | 75 | |
| BC171806.1 | - | 1 | 254 | 9054460 | 75 | |
| BC171885.1 | - | 1 | 317 | 9054539 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK093658.1, NM_001278283.1, NM_001318930.1, XM_006712677.3 transcripts.
Set Membership: |
|

RPE
Genomic Map