Assay Details
Gene Symbol: PCED1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271168.1 | NP_001258097.1 | 3 - 4 | 600 | 72 | |
XM_005260805.2 | XP_005260862.1 | 3 - 4 | 647 | 72 | ||
GenBank mRNA | AK026029.1 | - | 3 - 4 | 508 | 72 | |
BC014247.2 | - | 3 - 4 | 501 | 4641068 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | NM_022760.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | TC2000008200.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000356872.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000360652.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000439542.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000448755.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | ENST00000487501.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | NM_001271168.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000157787.hg.1 | NM_022760.4 | Refseq |
