Assay Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC011763.2 | - | 5 - 6 | 931 | 3617674 | 85 |
| BC025949.1 | - | 5 - 6 | 931 | 4620469 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201312.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201312.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201312.hg.1 | TC0700013594.hg.1 | Clariom D TC |

TRIM4
Genomic Map