Assay Details
| Gene Symbol: PTGER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028294.1 | 3 - 4 | 1333 | 98 | ||
| GenBank mRNA | X83863.1 | - | 3 - 4 | 1309 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002774.hg.1
Set Membership: |
|

PTGER3
Genomic Map