Assay Details
| Gene Symbol: PRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000947.4 | NP_000938.2 | 1 - 2 | 86 | 75 | |
| NM_001282487.1 | NP_001269416.1 | 1 - 2 | 86 | 75 | ||
| XM_017011004.1 | XP_016866493.1 | 1 - 2 | 86 | 75 | ||
| XM_017011005.1 | XP_016866494.1 | 1 - 2 | 86 | 75 | ||
| XM_017011006.1 | XP_016866495.1 | 1 - 2 | 86 | 75 | ||
| GenBank mRNA | AK223287.1 | - | 1 - 2 | 75 | 75 | |
| AK291915.1 | - | 1 - 2 | 73 | 75 | ||
| BC017833.1 | - | 1 - 2 | 86 | 75 | ||
| BC064931.1 | - | 1 - 2 | 71 | 6148494 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK291915.1, BC017833.1, BC064931.1, NM_000947.4, NM_001282487.1, XM_017011004.1, XM_017011005.1, XM_017011006.1 transcripts.
Set Membership: |
|

PRIM2
Genomic Map