Assay Details
| Gene Symbol: TSPAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308316.1 | NP_001295245.1 | 3 - 4 | 328 | 127 | |
| NM_005725.5 | NP_005716.2 | 3 - 4 | 328 | 127 | ||
| GenBank mRNA | AF054839.1 | - | 3 - 4 | 260 | 127 | |
| AK315385.1 | - | 3 - 4 | 288 | 127 | ||
| BC021675.1 | - | 3 - 4 | 293 | 4042036 | 127 | |
| BQ067895.1 | - | 3 - 4 | 340 | 127 | ||
| GU971730.1 | - | 3 - 4 | 260 | 127 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003017.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01047099.hg.1 | ENST00000369514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047099.hg.1 | ENST00000369515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047099.hg.1 | ENST00000369516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047099.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047099.hg.1 | TC01003017.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047100.hg.1 | ENST00000369514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047100.hg.1 | ENST00000369516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047100.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047100.hg.1 | TC01003017.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | ENST00000491992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015364.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | ENST00000369515.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | NM_001308315.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214838.hg.1 | TC0100015364.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | ENST00000369516.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | ENST00000433172.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | NM_001308316.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | NM_005725.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214839.hg.1 | TC0100015364.hg.1 | Clariom D TC |

TSPAN2
Genomic Map