Assay Details
| Gene Symbol: SYTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011496.1 | XP_016866985.1 | 4 - 5 | 1176 | 70 | |
| XM_017011497.1 | XP_016866986.1 | 5 - 6 | 513 | 70 | ||
| GenBank mRNA | AK304191.1 | - | 1 - 2 | 223 | 70 | |
| DB175440.1 | - | 5 - 6 | 514 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK304191.1, XM_017011496.1, XM_017011497.1 transcripts.
Set Membership: |
|

SYTL3
Genomic Map