Assay Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018471.2 | NP_060941.2 | 7 - 8 | 1095 | 67 | |
| XM_017004502.1 | XP_016859991.1 | 7 - 8 | 965 | 67 | ||
| GenBank mRNA | AF109366.1 | - | 7 - 8 | 954 | 67 | |
| AF161421.1 | - | 7 - 8 | 966 | 67 | ||
| AF220184.1 | - | 7 - 8 | 1096 | 67 | ||
| AF289586.1 | - | 7 - 8 | 995 | 67 | ||
| AK001901.1 | - | 7 - 8 | 1000 | 67 | ||
| AK297658.1 | - | 5 - 6 | 755 | 67 | ||
| AK298615.1 | - | 1049 | 67 | |||
| BC021102.1 | - | 7 - 8 | 907 | 5020890 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001098.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016456.hg.1 | ENST00000337859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016456.hg.1 | ENST00000536434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016456.hg.1 | ENST00000544130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016456.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016456.hg.1 | TC02001098.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171859.hg.1 | TC0200010214.hg.1 | Clariom D TC |

ZC3H15
Genomic Map