Assay Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018471.2 | NP_060941.2 | 8 - 9 | 1188 | 99 | |
| XM_017004502.1 | XP_016859991.1 | 8 - 9 | 1058 | 99 | ||
| GenBank mRNA | AF109366.1 | - | 8 - 9 | 1047 | 99 | |
| AF161421.1 | - | 8 - 9 | 1059 | 99 | ||
| AF220184.1 | - | 8 - 9 | 1189 | 99 | ||
| AK001901.1 | - | 8 - 9 | 1093 | 99 | ||
| AK298615.1 | - | 1142 | 99 | |||
| BC021102.1 | - | 8 - 9 | 1000 | 5020890 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001098.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016458.hg.1 | ENST00000337859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016458.hg.1 | ENST00000536434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016458.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016458.hg.1 | TC02001098.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000421536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000437396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000468120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000477672.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000481101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000496289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010214.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | ENST00000337859.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | ENST00000445547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | ENST00000498757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | NM_018471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171861.hg.1 | TC0200010214.hg.1 | Clariom D TC |

ZC3H15
Genomic Map